Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Opti-Bind™ Polystyrene Sulfate Particles

Achieve maximum reactivity in many diagnostic applications with Thermo Scientific™ Opti-Bind™ Polystyrene Sulfate Particles.

$304.00 - $936.00

Specifications

Certifications/Compliance Includes Certificate of Analysis
Content And Storage 2°C to 8°C
Description Opti-Bind Polystyrene Sulfate Particles
For Use With (Application) Diagnostic Assays
Material Polystyrene Sulfate
View More Specs

Products 16
Catalog Number Mfr. No. Diameter (Metric) Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Diameter (Metric) Quantity Price Quantity & Availability  
09-980-853
View Documents
Thermo Scientific™
71002697100250
1.25 μm 15 mL
Each for $347.00
Add to Cart
 
09-980-854
View Documents
Thermo Scientific™
71002697100350
1.25 μm 100 mL
Each for $936.00
Add to Cart
 
09-981-015
View Documents
Thermo Scientific™
71003497100250
2.5 μm 15 mL
Each for $304.00
Add to Cart
 
09-981-016
View Documents
Thermo Scientific™
71003497100350
2.5 μm 100 mL
Each for $925.00
Add to Cart
 
09-981-000
View Documents
Thermo Scientific™
81000397100290
0.1 μm 15 mL
Each for $347.00
Add to Cart
 
09-981-001
View Documents
Thermo Scientific™
81000397100390
0.1 μm 100 mL
Each for $936.00
Add to Cart
 
09-981-003
View Documents
Thermo Scientific™
81000597100290
0.2 μm 15 mL
Each for $347.00
Add to Cart
 
09-981-004
View Documents
Thermo Scientific™
81000597100390
0.2 μm 100 mL
Each for $936.00
Add to Cart
 
09-981-006
View Documents
Thermo Scientific™
81000797100290
0.3 μm 15 mL
Each for $347.00
Add to Cart
 
09-981-007
View Documents
Thermo Scientific™
81000797100390
0.3 μm 100 mL
Each for $936.00
Add to Cart
 
09-980-855
View Documents
Thermo Scientific™
81000997100290
0.4 μm 15 mL
Each for $347.00
Add to Cart
 
09-980-856
View Documents
Thermo Scientific™
81000997100390
0.4 μm 100 mL
Each for $936.00
Add to Cart
 
09-980-841
View Documents
Thermo Scientific™
91001397100290
0.6 μm 15 mL
Each for $347.00
Add to Cart
 
09-980-842
View Documents
Thermo Scientific™
91001397100390
0.6 μm 100 mL
Each for $936.00
Add to Cart
 
09-980-857
View Documents
Thermo Scientific™
91001897100290
0.85 μm 15 mL
Each for $347.00
Add to Cart
 
09-980-858
View Documents
Thermo Scientific™
91001897100390
0.85 μm 100 mL
Each for $936.00
Add to Cart
 
Description

Description

Achieve maximum reactivity in many diagnostic applications with Thermo Scientific™ Opti-Bind™ Polystyrene Sulfate Particles for use in turbidimetric/nephelometric assays, slide agglutination assays, chemiluminescent assays and in clinical diagnostics and molecular biology applications. Particles can be used direct from the bottle, without any pre-cleaning.

Our production does not utilize common surfactants (like SDS, Tween™ 20, Triton™ X-100) that can interfere with protein binding to particle surfaces. Polystyrene surfaces are very hydrophobic and adsorb proteins almost instantaneously. Our proprietary anionic surfactant does not interfere with the binding of proteins or cause proteins to desorb from microparticle surfaces.
  • Available in a wide range of diameters (from 0.1 μM to 2.5 μM)
  • No pre-washing
  • Tight size distribution—CV<2%
  • Optimized colloidal stability
  • Shelf-life of 10 years

Recommended for:

Clinical diagnostics, molecular biology, nephelometric, turbidimetric and slide agglutination assays

Note:

When using Opti-Bind™ particles, ask about our Microparticle Reagent Optimization manual, which outlines our reagent development strategy. This manual includes validated binding, coupling and particle reagent optimization protocols.

Specifications

Specifications

Includes Certificate of Analysis
Opti-Bind Polystyrene Sulfate Particles
Polystyrene Sulfate
Package Insert and Certificate of Analysis
2°C to 8°C
Diagnostic Assays
Specialty Particles
Opti-Bind Polystyrene particles are available in different concentrations and package sizes upon request.
Safety and Handling

Safety and Handling

ShelfLife : 10 Years

SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit