Learn More
Thermo Scientific™ Sensors and Replacement Batteries for Thermo Scientific™ Ultra-Low Temperature Freezers
Use these Thermo Scientific™ Sensors and Replacement Batteries with our ultra-low temperature freezers.
Manufacturer:  Thermo Scientific™ 290199
 View more versions of this product
Catalog No. 29-019-9
  Check Availability
Add to cart
Specifications
Description
Use these Thermo Scientific™ Sensors and Replacement Batteries with our ultra-low temperature freezers.
Description & Specifications
Specifications
Optional PT100 sensor for connection to building automation systems (Factory installed)
Forma 900 and 7000 Series, HERAfreeze HFU B Series, Revco ExF and DxF Series, and TSE and TSD Series
Sensors and Replacement Batteries
Forma™ 7000