Learn More
OptiVISOR™
Manufacturer:  Moore Medical Corp 89081
 View more versions of this product
Catalog No. 19-898-851
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
For unrestricted efficiency while reducing eye strain, leaves both hands free, allows three dimensional vision, used by anyone whose profession or craft requires close accurate work
Prescription or safety eyeglasses
2.75x
Binocular Headband Magnifier