OptiVISOR™
Manufacturer: Moore Medical Corp 97655
 View more versions of this product
Catalog No. 19-899-047
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) For unrestricted efficiency while reducing eye strain, leaves both hands free, allows three dimensional vision, used by anyone whose profession or craft requires close accurate work
    For Use With (Equipment) Prescription or safety eyeglasses
    Magnification 1.75x
    Product Type Binocular Headband Magnifier
  • Description & Specifications
Related Products
Product Content Feedback