Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ Alkaline Phosphatase, Fisher BioReagents™

 View more versions of this product

Catalog No. BP80971


Order in Bulk

Request bulk or custom formats

Add to cart

Description

Description

OPTIZYME™ AP is a novel alkaline phosphatase that catalyzes the release of 5'- and 3'-phosphate groups from DNA, RNA and nucleotides in 10min.

Applications and Features:

  • OPTIZYME™ Alkaline Phosphatase dephosphorylates cloning vector DNA (10min at 37°C)
  • Fast thermoinactivation (5min at 75°C)
  • Simultaneous digestion and dephosphorylation of vector DNA
  • Single protocol for all types of DNA ends
  • Nucleotide degradation prior to sequencing PCR products
  • Active on nucleotides
  • Protein dephosphorylation
  • Active in all OPTIZYME restriction enzyme buffers

Supplied with:
10X OPTIZYME AP Buffer: 100mM Tris-HCl (pH 8.0 at 37°C), 50mM MgCl2, 1M KCl, 0.2% (v/v) Triton X-100, 1mg/mL BSA
Source: E. Coli cells with a cloned bacterial alkaline phosphatase gene
Quality Control:

  • Endo-, exodeoxyribonuclease and ribonuclease-free
  • Functionally tested for dephosphorylation of 5'-termini of overhanging, recessed and blunt DNA

Specifications

Specifications

Alkaline Phosphatase
1U/μL
E. coli cells with a cloned bacterial Alkaline Phosphatase gene
1000U
OPTIZYME™ AP Buffer 10X : Tris HCl 100mM (8 pH at 37°C), MgCl2 50mM, KCl 1M, Triton™ X-100 0.2%, BSA 1mg/mL
Solid
Colorless
7.4
DePhosphorylates cloning vector DNA (10 min. at 37°C), fast thermo-inactivation (5 min. at 75°C), Nucleotide degradation prior to sequencing PCR products, active on Nucleotides, Protein dephosphorylation, restriction En
-20°C
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit