Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ DNA Polymerase I, Fisher BioReagents™

$351.50

Specifications

Color Colorless
Concentration 10U/μL
Product Type DNA Polymerase I
Format Liquid
For Use With (Application) DNA Polymerase I labels DNA by nick translation in conjunction with DNase I, second-strand cDNA synthesis in conjunction with RNase H, incorporation of modified Nucleotides, Reverse Transcriptases
View More Specs

Products 2
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
BP81095
View Documents
2,500U Each for $351.50
Order in Bulk

Request bulk or custom formats

Add to cart
 
BP81091
View Documents
500U This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

OPTIZYME™ DNA Pol I is a template-dependent DNA Polymerase that catalyzes 5'->3' DNA synthesis, and exhibits 3'->5'-exonuclease, 5'->3' exonuclease and RNase H activity.

Applications and Features:

  • OPTIZYME DNA Polymerase I labels DNA by nick translation in conjunction with DNase I.
  • Second-strand cDNA synthesis in conjunction with RNase H.
  • Incorporation of modified nucleotides (e.g., biotin-, digoxigenin-, aminoallyl-, fluorescently-labeled nucleotides).
  • Active in buffers for OPTIZYME restriction enzymes and reverse transcriptases.

Supplied with:
10X OPTIZYME DNA Pol I Buffer: 500mM Tris-HCl (pH 7.5 at 25°C), 100mM MgCl2, 10mM DTT

Source: E. Coli cells with a cloned polA gene for E.Coli

Quality Control:
Endodeoxyribonuclease-free

Specifications

Specifications

Colorless
DNA Polymerase I
DNA Polymerase I labels DNA by nick translation in conjunction with DNase I, second-strand cDNA synthesis in conjunction with RNase H, incorporation of modified Nucleotides, Reverse Transcriptases
E. coli cells with a cloned polA gene
-20°C
10U/μL
Liquid
7.5
OPTIZYME™ DNA Pol I Buffer 10X : Tris HCl 500mM (7.5 pH at 25°C), MgCl2 100mM, DTT 10mM
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit