Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Description
5'...G^A A T T C...3'
3'...C T T A A^G...5'
Conditions for 100% Activity:
- 1X OPTIZYME Buffer EcoRI: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2, 100mM NaCl, 0.02% Triton X-100 and 0.1mg/ml BSA.
- Incubate at 37°C.
Enzyme Activity in OPTIZYME buffers:
- Buffer 1: 0 - 20%
- Buffer 2: No reaction
- Buffer 3: 100%
- Buffer 4: No reaction
- Buffer 5: 100%™
Storage Buffer:
10mM potassium phosphate (pH 7.4 at 25°C), 300mM NaCl, 1mM EDTA, 1mM DTT, 0.2mg/ml BSA, 0.15% Triton X-100 and 50% (v/v) glycerol.
Ligation and Recleavage:
More than 95% of DNA fragments can be ligated and recut after a 50-fold overdigestion with EcoRI.
Digestion of Agarose-embedded DNA:
A minimum of 5 units of EcoRI is required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
Note: Low salt concentration, large excess of the enzyme, pH>8.0, or the replacement of mg2+ by Mn2+ may result in star activity.
Compatible Ends: XapI, MunI, TasI.
Methylation Effects:
Dam: never overlaps - no effect.
Dcm: never overlaps - no effect.
CpG: may overlap - cleavage impaired.
- 5'...m5C GAATTm5C G...3'
- 5'...TGm6AATTC(N)4TGCT...3'
Specifications
10 U/μL | |
37°C | |
>90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with EcoRI | |
Keep container tightly closed | |
G.AATTC | |
Liquid |
25 to 50% Water, >50% Glycerin, 2.5 to10% Sodium Chloride | |
7.4 | |
10mM Potassium Phosphate (pH 7.4 at 25°C), 300mM NaCl, 1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 0.15% Triton™ X-100, 50% Glycerol | |
7500 U | |
EcoRI |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction