Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ Klenow Fragment, Fisher BioReagents™

Specifications

Color Colorless
Concentration 10U/μL
Product Type Klenow Fragment
Format Liquid
For Use With (Application) Klenow fragment blunts DNA ends by filling in 5'-overhangs, random-primed DNA labeling, DNA sequencing by the sanger method, second strand synthesis of cDNA, site-specific mutagenesis of DNA with synthetic Oligonucleotides, lacks the 5'->3' Exonuclease activity of DNA Polymerase I
View More Specs

Products 2
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
BP81061
View Documents
300U This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
BP81065
View Documents
1500U This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

OPTIZYME™ Klenow Fragment is the large fragment of DNA Pol I and exhibits both 5'->3' polymerase and 3'->5' exonuclease activity.

Applications and Features:

  • OPTIZYME Klenow Fragment blunts DNA ends by filling in 5'-overhangs
  • Random-primed DNA labeling
  • Labeling by filling in 5'-overhangs of double-stranded DNA
  • Random-primed DNA labeling
  • DNA sequencing by the Sanger Method
  • Second strand synthesis of cDNA
  • Site-specific mutagenesis of DNA with synthetic oligonucleotides
  • Active in buffers for OPTIZYME restriction enzymes, reverse transcriptases and T4 DNA ligase
  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, aminoallyl, biotin-, digoxigenin-, and fluorescently-labeled nucleotides)
  • Lacks the 5'->3' exonuclease activity of DNA Polymerase I

Supplied with:
10X OPTIZYME Klenow Fragment Buffer: 500mM Tris-HCl (pH 8.0 at 25°C), 50mM MgCl2, 10mM DTT
Source: E. Coli cells with a cloned fragment of the polA gene
Quality Control:

  • Endodeoxyribonuclease-free
  • Functionally tested for fill in of 5'-overhanging DNA termini and for random primed DNA labeling

Specifications

Specifications

Colorless
Klenow Fragment
Klenow fragment blunts DNA ends by filling in 5'-overhangs, random-primed DNA labeling, DNA sequencing by the sanger method, second strand synthesis of cDNA, site-specific mutagenesis of DNA with synthetic Oligonucleotides, lacks the 5'->3' Exonuclease activity of DNA Polymerase I
E. coli cells with a cloned fragment of the polA gene
-20°C
10U/μL
Liquid
7.5
OPTIZYME™ Klenow Fragment Buffer 10X : Tris HCl 500mM (8 pH at 25°C), MgCl2 50mM, DTT 10mM
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit