Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ M-MLV Reverse Transcriptase, Fisher BioReagents™

$68.10 - $202.00

Specifications

Color Colorless
Concentration 200U/μL
Product Type OPTIZYME™ M-MLV Reverse Transcriptase
Format Liquid
For Use With (Application) Generation of labeled cDNA probes for microarrays, synthesis of cDNA for cloning and expression, analysis of RNA by primer extension, DNA labeling, optimum activity at 42°C, incorporates modified Nucleotides
View More Specs

Products 2
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
BP81041
View Documents
5,000U Each for $68.10
Order in Bulk

Request bulk or custom formats

Add to cart
 
BP81045
View Documents
25,000U Each for $202.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
Description

Description

OPTIZYME™ M-MuLV RT is a genetically modified MLV RT that possesses RNA- and DNA-dependent polymerase activity.

Applications and Features:

  • OPTIZYME M-MLV RT is used for efficient first strand cDNA synthesis for RT-PCR and real-time RT-PCR (up to 13kb)
  • Generation of labeled cDNA probes for microarrays
  • Synthesis of cDNA for cloning and expression
  • Analysis of RNA by primer extension
  • DNA labeling
  • Optimum activity at 42°C (active up to 50°C)
  • Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, fluorescein-labeled nucleotides)

Supplied with:
5X OPTIZYME M-MLV RT Buffer: 250mM Tris-HCl (pH 8.3 at 25°C), 250mM KCl, 20mM MgCl2, 50mM DTT
Source: E. Coli cells with a cloned fragment of the pol gene encoding Moloney Murine Leukemia Virus reverse transcriptase
Quality Control:

  • Endo-, exodeoxyribonuclease, phosphatase and ribonuclease-free
  • Functionally tested in first strand cDNA and RT-PCR

Specifications

Specifications

Colorless
OPTIZYME™ M-MLV Reverse Transcriptase
Generation of labeled cDNA probes for microarrays, synthesis of cDNA for cloning and expression, analysis of RNA by primer extension, DNA labeling, optimum activity at 42°C, incorporates modified Nucleotides
E. coli cells with a cloned fragment of the pol gene encoding Moloney Murine Leukemia Virus Reverse Transcriptase
-20°C
200U/μL
Liquid
8.3
OPTIZYME™ M-MLV RT Buffer 5X : Tris HCl 250mM (8.3 pH at 25°C), KCl 250mM, MgCl2 20mM, DTT 50mM
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit