Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OPTIZYME™ NotI, Fisher BioReagents™
Specifications
Concentration | 10 U/μL |
---|---|
Components | 25 to 50% Water, >50% Glycerin |
pH | 7.4 |
For Use With (Application) | >90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with Not I |
Storage Buffer | 20mM Tris HCl (pH 7.8 at 25°C), 100mM NaCl, 0.1mM EDTA, 1mM DTT, 0.02% Triton™ X-100, 0.2mg/mL BSA, 50% Glycerol |
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Quantity | Price | Quantity & Availability | ||||||
BP8004-5
|
1000 U | Item Discontinued This item has been discontinued by Fisher Scientific. Please Sign In to view product availability in your area. | N/A | ||||||
BP8004-1
|
300 U | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||||||
Description
5'...G C^G G C C G C...3'
3'...C G C C G G^C G...5'
Conditions for 100% Activity
- 1X OPTIZYME Buffer 3: 50mM Tris-HCl (pH 7.5 at 37°C), 10mM MgCl2 , 100mM NaCl and 0.1mg/ml BSA.
- Incubate at 37°C.
Enzyme Activity in OPTIZYME buffers
- Buffer 1: 0 - 20%
- Buffer 2: 20 - 50%
- Buffer 3: 100%
- Buffer 4: 0 - 20%
- Buffer 5: 20 - 50%
20mM Tris-HCl (pH 7.8 at 25°C), 100mM NaCl, 0.1mM EDTA, 1mM DTT, 0.02% Triton X-100, 0.2mg/ml BSA and 50% (v/v) glycerol.
Ligation and Recleavage:
More than 95% of DNA fragments can be ligated and recut after a 50-fold overdigestion with NotI.
Digestion of Agarose-embedded DNA:
A minimum of 5 units of NotI is required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
Compatible Ends: Bsp120I, CfrI, Eco52I.
Methylation Effects:
Dam: never overlaps - no effect.
Dcm: never overlaps - no effect.
CpG: completely overlaps - blocked.
- GCGGCCGC
Specifications
10 U/μL | |
7.4 | |
20mM Tris HCl (pH 7.8 at 25°C), 100mM NaCl, 0.1mM EDTA, 1mM DTT, 0.02% Triton™ X-100, 0.2mg/mL BSA, 50% Glycerol | |
GC.GGCCGC | |
Liquid |
25 to 50% Water, >50% Glycerin | |
>90% of DNA fragments can be ligated and re-cut after 50-fold over-digestion with Not I | |
Keep container tightly closed | |
NotI |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title