Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ ScaI, Fisher BioReagents™

Catalog No. BP8037-1

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

5'...A G T^A C T...3'
3'...T C A^T G A...5'

Supplied with: 10X OPTIZYME AarI, ScaI, PasI Buffer
Conditions for 100% Activity:

  • 1X OPTIZYME Buffer AarI, ScaI, PasI: 10mM Bis-Tris Propane-HCl (pH 6.5 at 37°C), 10mM MgCl2 , 100mM KCl and 0.1mg/ml BSA
  • Incubate at 37°C
Enzyme Activity in OPTIZYME buffers:
  • Buffer 1: 0 - 20%
  • Buffer 2: 0 - 20%
  • Buffer 3: 0 - 20%
  • Buffer 4: 0 - 20%
  • Buffer 5: 0 - 20%
Storage Buffer:
  • 10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.5mg/ml BSA and 50% (v/v) glycerol
  • Ligation and Re-cleavage:
  • More than 80% of the DNA fragments can be ligated and more than 90% of these can be re-cut after 50-fold over-digestion with ScaI.
  • Digestion of Agarose-embedded DNA:
  • A minimum of 20 units of ScaI is required for the complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours.
  • Notes:
    • Conditions of low salt, high enzyme concentration, glycerol concentration >5% or pH >8.0 may result in star activity.
    • Supercoiled plasmids may require up to 20-fold more ScaI for complete digestion than linear DNA.
    Methylation Effects:
    Dam: Never overlaps - no effect
    Dcm: Never overlaps - no effect
    CpG: Never overlaps - no effect
    EcoKI: Never overlaps - no effect
    EcoBI: May overlap - blocked
    • 5'...TGm6AGTACT(N)3 TGCT...3'
  • 3'...AC TCATGA(N)3 m6ACGA...5'
  • (Cleavage blocked)
  • Specifications

    Specifications

    ScaI
    10U/μL
    10X OPTIZYME™ AarI, ScaI, PasI Buffer
    7.4
    >80% of DNA fragments can be ligated and >90% of these can be re-cut after 50-fold over-digestion with ScaI
    Liquid
    AGT.ACT
    37°C
    1000U
    10mM Tris HCl (pH 7.4 at 25°C), 100mM KCl, 1mM EDTA, 1mM DTT, 0.2mg/mL BSA, 50% Glycerol
    SDS
    Documents
    Product Certifications
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit