Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ T3 RNA Polymerase, Fisher BioReagents™

 View more versions of this product

Catalog No. BP81015

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

OPTIZYME™ T3 RNA Pol is a DNA-dependent RNA polymerase that catalyzes the 5'->3' synthesis of RNA from DNA.

Applications and Features:

  • OPTIZYME™ T3 RNA Polymerase synthesizes unlabeled and labeled RNA that can be used for: (1) hybridization, (2) in vitro RNA translation, (3) RNase protection assays, (4) genomic DNA sequencing, (5) RNA splicing, and (6) studies of RNA secondary structure and RNA-protein interactions
  • Incorporation of modified nucleotides (e.g., aminoallyl-, biotin-, fluorescein-, digoxigenin-labeled nucleotides)

Supplied with:
5X OPTIZYME Transcription Buffer: 200mM Tris-HCl (pH 7.9 at 25°C), 30mM MgCl2, 50mM DTT, 50mM NaCl, 10mM spermidine
Source: E. Coli cells with a cloned T3 RNA Polymerase gene
Quality control:

  • Endo-, exodeoxyribonuclease and ribonuclease-free
  • Functionally tested in in vitro transcription reaction

Specifications

Specifications

T3 RNA Polymerase
20U/μL
E. coli cells with a cloned T3 RNA Polymerase gene
5,000U
OPTIZYME™ Transcription Buffer 5X : Tris HCl 200mM (7.9 pH at 25°C), MgCl2 30mM, DTT 50mM, NaCl 50mM, Spermidine 10mM
Liquid
Colorless
7.5
Hybridization, in-vitro RNA translation, RNase protection assay, genomic DNA sequencing, RNA splicing and studies of RNA secondary structure and RNA-Protein interaction, quality control
-20°C
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit