Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ T4 Polynucleotide Kinase, Fisher BioReagents™

$324.00

Specifications

Color Colorless
Concentration 10U/μL
Product Type T4 Polynucleotide Kinase
Format Liquid
For Use With (Application) Probes for hybridization or transcript mapping, primers for PCR or DNA sequencing and markers for Gel electrophoresis, removal of 3'-phosphate groups
View More Specs

Products 2
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
BP80981
View Documents
500U
Each for $324.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
BP80985
View Documents
2,500U This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

OPTIZYME™ T4 PNK catalyzes the transfer of γ-phosphate from ATP to the 5'-OH group of DNA, RNA, oligonucleotides and nucleoside 3'-monophosphates.

Applications and Features:

  • OPTIZYME™ T4 PNK labels 5'-termini of nucleic acids (for use as: probes for hybridization or transcript mapping, primers for PCR or DNA sequencing, and markers for gel-electrophoresis)
  • 5'-phosphorylation of oligonucleotides, PCR products, and other DNA or RNA prior to ligation
  • Phosphorylation of PCR primers
  • Removal of 3'-phosphate groups
  • Active in buffers for OPTIZYME restriction enzymes, T4 DNA Ligase and Reverse Transcriptases

Supplied With:

  • 10X OPTIZYME T4 PNK Buffer 1 (for forward reaction): 500mM Tris-HCl (pH7.6 at 25°C), 100mM MgCl2, 50mM DTT, 1mM spermidine
  • 10X OPTIZYME T4 PNK Buffer 2 (for exchange reaction): 500mM imidazole-HCl (pH6.4 at 25°C), 180mM MgCl2, 50mM DTT, 1mM spermidine, 1mM ADP
  • 24% PEG 6000 Sol for T4 PNK: 24% (w/v) polyethylene glycol 6000
Source: E. Coli cells with a cloned pse T gene of bacteriophage T4
Quality Control:
  • Endo-, exodeoxyribonuclease and ribonuclease-free
  • Functionally tested for labeling of 5'-termini of DNA

Specifications

Specifications

Colorless
T4 Polynucleotide Kinase
Probes for hybridization or transcript mapping, primers for PCR or DNA sequencing and markers for Gel electrophoresis, removal of 3'-phosphate groups
E. coli cells with a cloned pse T gene of bacteriophage T4
-20°C
10U/μL
Liquid
7.5
10X OPTIZYME™ T4 PNK Buffer 1 (for forward reaction): 500mM Tris HCl (7.6 pH at 25°C), 100mM MgCl2, 50mM DTT, 1mM spermidine, 10X OPTIZYME™ T4 PNK Buffer 2 (for exchange reaction): 500mM imidazole-HCl
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit