Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OPTIZYME™ Terminal Deoxynucleotidyl Transferase, Fisher BioReagents™

$218.00

Specifications

Color Colorless
Concentration 20U/μL
Product Type Terminal Deoxynucleotidyl Transferase
Format Liquid
For Use With (Application) Homopolymeric tailing of linear duplex DNA with any type of 3'-OH terminus, Oligodeoxyribonucleotide and DNA labeling, 5'-rapid amplification of cDNA Ends, in-situ localization of apoptosis incorporation of modified Nucleotides
View More Specs

Products 2
Catalog Number Quantity Price Quantity & Availability  
Catalog Number Quantity Price Quantity & Availability  
BP81035
View Documents
1500U
Each for $218.00
Order in Bulk

Request bulk or custom formats

Add to cart
 
BP81031
View Documents
300U This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

OPTIZYME™ TdT is a template-independent DNA polymerase that catalyzes the repetitive addition of deoxyribonucleotides to the 3'-OH group of DNA

Applications and Features:

  • OPTIZYME TdT produces synthetic homo- and heteropolymers
  • Homopolymeric tailing of linear duplex DNA with any type of 3'-OH terminus
  • Oligodeoxyribonucleotide and DNA labeling
  • 5'-Rapid Amplification of cDNA Ends (5'-RACE)
  • In situ localization of apoptosis
  • Incorporation of modified nucleotides (e.g., fluorescein-, biotin-, aminoallyl-labeled nucleotides)

Supplied with:
5X OPTIZYME TdT Buffer: 1M potassium cacodylate, 125mM Tris, 0.05% (v/v) Triton X-100, 5mM CoCl2 (pH 7.2 at 25°C)

Source: E. Coli cells with a cloned gene encoding calf thymus terminal deoxynucleotidyl transferase

Quality Control:
Endo-, exodeoxyribonuclease, phosphatase and ribonuclease-free

Specifications

Specifications

Colorless
Terminal Deoxynucleotidyl Transferase
Homopolymeric tailing of linear duplex DNA with any type of 3'-OH terminus, Oligodeoxyribonucleotide and DNA labeling, 5'-rapid amplification of cDNA Ends, in-situ localization of apoptosis incorporation of modified Nucleotides
E. coli cells with a cloned gene encoding calf thymus terminal deoxynucleotidyl transferase
-20°C
20U/μL
Liquid
7.5
OPTIZYME™ TdT Buffer 5X : Potassium cacodylate 1M, Tris 125mM, Triton™ X-100 0.05%, CoCl2 5mM (7.2 pH at 25°C)
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit