Learn More
Aviva Systems Biology Corp OPTN ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP32292_P050
This is a rabbit polyclonal antibody against OPTN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human OPTN. Alternate names: FIP2, GLC1E, HIP7, HYPL, NRP, TFIIIA-INTP, ALS12ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54513