Learn More
Tocris Bioscience™ OptoBI-1
Photoswitchable TRPC3/6/7 agonist
Manufacturer:  Tocris Bioscience™ 7013/10
 View more versions of this product
Catalog No. 70-131-0
Add to cart
This item is not returnable. View return policy
Description
Description
OptoBI-1 is a photoswitchable TRPC3/6/7 agonist (EC50 for active cis-form ∽0.1 μM at TRPC3). Switches conformation from trans to cis at 365 nm and cis to trans at 430 nm. Returns to trans conformation in the dark after ∽ 50 minutes. No effect observed on TRPC4 or 5. Rapidly changes TRPC3 conductance and inhibits neuronal firing in cis-state.
Specifications
Specifications
1-[1-[4-[4-[(4-Butylphenyl)diazenyl]phenyl]butanoyl]piperidin-4-yl]-1,3-dihydro-2H-benzo[d]imidazol-2-one
C32H37N5O2
98%
10 mg