Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ OptoBI-1

Photoswitchable TRPC3/6/7 agonist

$209.00 - $881.00

Specifications

Chemical Name or Material 1-[1-[4-[4-[(4-Butylphenyl)diazenyl]phenyl]butanoyl]piperidin-4-yl]-1,3-dihydro-2H-benzo[d]imidazol-2-one
Molecular Formula C32H37N5O2
Purity 98%
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
70-131-0
View Documents
Tocris Bioscience™
7013/10
10 mg
Each for $209.00
Add to cart
 
70-135-0
View Documents
Tocris Bioscience™
7013/50
50 mg
Each for $881.00
Add to cart
 
Description

Description

  • Photoswitchable TRPC3/6/7 agonist (EC50 for active cis-form ∼0.1 μM at TRPC3).
  • Switches conformation from trans to cis at 365 nm and cis to trans at 430 nm.
  • Returns to trans conformation i the dark after ∼ 50 minutes.
  • No effect observed on TRPC4 or 5.
  • Rapidly changes TRPC3 conductance and inhibits neuronal firing in cis-state.
Specifications

Specifications

1-[1-[4-[4-[(4-Butylphenyl)diazenyl]phenyl]butanoyl]piperidin-4-yl]-1,3-dihydro-2H-benzo[d]imidazol-2-one
C32H37N5O2
98%
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit