Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR10A5 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS11631R

Description
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.Specifications
OR10A5 | |
Polyclonal | |
Unconjugated | |
OR10A5 | |
HP3; JCG6; Olfactory receptor 10A1; Olfactory receptor 10A5; Olfactory receptor 11-403; olfactory receptor family 10 subfamily A member 5; olfactory receptor OR11-85; olfactory receptor, family 10, subfamily A, member 1; olfactory receptor, family 10, subfamily A, member 5; Olfactory receptor-like protein JCG6; OR10A1; OR10A5; OR11-403 | |
Rabbit | |
Protein A | |
RUO | |
144124 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q9H207 | |
OR10A5 | |
KLH conjugated synthetic peptide derived from human OR10A5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction