Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR10R2 Polyclonal Antibody
Rabbit Polyclonal Antibody
$589.50
Specifications
Antigen | OR10R2 |
---|---|
Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5111495
|
Invitrogen™
PA5111495 |
100 μL |
Each for $589.50
|
|
|||||
PIPA5111503
|
Invitrogen™
PA5111503 |
100 μL |
Each for $589.50
|
|
|||||
Description
Immunogen sequence: RNKDVQLAIR KVLGKKGSLK LYN.
G-protein Coupled Receptors (GPCRs) comprise one of the largest families of signaling molecules with more thana thousandmembers currently predicted to exist.All GPCRs share a structural motif consisting of seven membrane-spanning helices, and exist in both active and inactive forms. An array of activating ligands participate in the conformation of GPCRs which leads to signaling via G-proteins and downstream effectors. Ongoing studies have also shown the vast series of reactions which participate in the negative regulation of GPCRs. This turn-off activity has tremendous implications for the physiological action of the cell, and continues to drive pharmacological research for new drug candidates.Specifications
OR10R2 | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NGX6 | |
343406 | |
Recombinant Protein corresonding to Human OR10R2. Recombinant protein control fragment (Product #RP-101605). | |
Antibody |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
OR10R2 | |
Olfactory receptor 10R2; olfactory receptor family 10 subfamily R member 2; olfactory receptor OR1-8; olfactory receptor, family 10, subfamily R, member 2; OR10R2; OR10R2Q; OR1-8 | |
OR10R2 | |
Primary | |
Antigen affinity chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title