Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR11H4 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA572250
Description
Putative odorant receptor. Multi-pass membrane protein. Belongs to the G-protein coupled receptor 1 family.
Specifications
OR11H4 | |
Polyclonal | |
Unconjugated | |
OR11H4 | |
Olfactory receptor 11H4; Olfactory receptor OR14-36; OR11H4 | |
Rabbit | |
Protein A | |
RUO | |
390442 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.31 mg/mL | |
PBS with 0.09% sodium azide | |
Q8NGC9 | |
OR11H4 | |
KLH conjugated synthetic peptide between 239-271 amino acids from the C-terminal region of human OR11H4 | |
80 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction