Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR14J1 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548706
Description
Odorant receptor.
Specifications
OR14J1 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q9UGF5 | |
OR14J1 | |
KLH conjugated synthetic peptide between 206-234 amino acids from the C-terminal region of human OR14J1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
OR14J1 | |
Olfactory receptor 14J1, Hs6M1-28, Olfactory receptor 5U1, Olfactory receptor OR6-25, OR14J1, OR5U1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
442191 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction