Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR1G1 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$482.50

Specifications

Antigen OR1G1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP168970
SDP
View Documents
Novus Biologicals
NBP168970
100 μL
Each of 1 for $482.50
Add to Cart
 
Description

Description

OR1G1 Polyclonal specifically detects OR1G1 in Human samples. It is validated for Western Blot.
Specifications

Specifications

OR1G1
Polyclonal
Rabbit
P47890
8390
Synthetic peptides corresponding to OR1G1 (olfactory receptor, family 1, subfamily G, member 1) The peptide sequence was selected from the C terminal of OR1G1. Peptide sequence FSSPSTHSAQKDTVASVMYTVVTPMLNPFIYSLRNQEIKSSLRKLIWVRK.
Primary
Western Blot
Unconjugated
RUO
Olfactory receptor 17-209, olfactory receptor 1G1, Olfactory receptor 1G2, Olfactory receptor OR17-8, olfactory receptor, family 1, subfamily G, member 1, olfactory receptor, family 1, subfamily G, member 2, OR17-209OR17-130, OR1G2
OR1G1
IgG
35 kDa
SDS
Documents

Documents

Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit