Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR1N1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309762100UL
Description
OR1N1 Polyclonal specifically detects OR1N1 in Human samples. It is validated for Western Blot.Specifications
OR1N1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Olfactory receptor 1-26, olfactory receptor 1N1, Olfactory receptor 1N3, Olfactory receptor OR9-22, olfactory receptor, family 1, subfamily N, member 1, olfactory receptor, family 1, subfamily N, member 3, OR1-26OR9-22, OR1N3 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR1N1 (NP_036495). Peptide sequence PPSIASEEKDIAAAAMYTIVTPMLNPFIYSLRNKDMKGALKRLFSHRSIV | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
138883 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction