Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR1S1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP168969
Description
OR1S1 Polyclonal specifically detects OR1S1 in Human samples. It is validated for Western Blot.Specifications
OR1S1 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
Q8NGQ3 | |
OR1S1 | |
Synthetic peptides corresponding to OR1S1 (olfactory receptor, family 1, subfamily S, member 1) The peptide sequence was selected from the C terminal of OR1S1. Peptide sequence FPSSTHPEDTDKIGAVLFTVVTPMINPFIYSLRNKDMKGALRKLINRKIS. | |
Affinity Purified | |
RUO | |
219959 | |
Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Western Blot | |
Unconjugated | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
olfactory receptor 1S1, Olfactory receptor OR11-232, olfactory receptor, family 1, subfamily S, member 1, OR11-232, OST034 | |
Rabbit | |
37 kDa | |
100 μL | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title