Learn More
OR2A2 Mouse anti-Human, Clone: 4B10, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA524541
Catalog No. PIMA524541
Add to cart
Description
Description
Peptide Sequence: PDSNQREEQE KMLSLFHSVL NPMLNPLIYS LRNAQLKGAL HRALQRKRSM RTVYGLCL
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
Specifications
OR2A2
Monoclonal
Unconjugated
OR2A2
Liquid
OR2A2
OR2A2 (NP_001005480.1, 261 a.a. ∽ 318 a.a) partial recombit protein with GST tag. MW of the GST tag alone is 26 KDa.
Protein A
RUO
Antibody
Monoclonal
Human
ELISA
4B10
PBS with no preservative; pH 7.4
Q6IF42
OR2A2; OR2A17P; OR2A2P
Mouse
IgG2a, kappa
100 μg
-20° C
Primary
442361