Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR2A5/OR2A14/OR2A42 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific OSR00272W

Catalog No. PIOSR00272W

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

Reconstitute with 100 μL of distilled water.

Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OR2A5/OR2A14/OR2A42
Polyclonal
OR2A14
Olfactory receptor 2A5, Olfactory receptor 7-138/7-141, OR7-138, OR7-141, OR2A26, OR2A8, Olfactory receptor 2A14, Olfactory receptor OR7-12, OST182, O
Rabbit
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q8NGT9, Q96R47, Q96R48
OR2A1, OR2A14, OR2A5
A synthetic peptide from human olfactory receptor
RUO
135941, 346528, 393046
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit