Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR2B11 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548198
Description
Odorant receptor.
Specifications
OR2B11 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q5JQS5 | |
OR2B11 | |
KLH conjugated synthetic peptide between 248-276 amino acids from the C-terminal region of human OR2B11. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
OR2B11 | |
Olfactory receptor 2B11, OR2B11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
127623 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction