Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR2M2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309772100UL
Description
OR2M2 Polyclonal specifically detects OR2M2 in Human samples. It is validated for Western Blot.Specifications
OR2M2 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
olfactory receptor 2M2, olfactory receptor, family 2, subfamily M, member 2, OST423OR2M2Q | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR2M2 (NP_001004688). Peptide sequence DKMVSVFYTILTPMLNPLIYSLRNKEVTRAFMKILGKGKSESELPHKLYV | |
100 μg | |
Primary | |
Human | |
Purified |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
391194 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction