Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR2V2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548547
Description
Odorant receptor.
Specifications
OR2V2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
Q96R30 | |
OR2V2 | |
KLH conjugated synthetic peptide between 245-273 amino acids from the C-terminal region of human OR2V2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
OR2V2 | |
Olfactory receptor 2V2, Olfactory receptor 2V3, Olfactory receptor OR5-3, OR2V2, OR2V3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
285659 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction