Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR2W3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561043
Description
Immunogen sequence: GHHEVDHFLR EMPALIRMAC VSTVAIE Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.
Specifications
OR2W3 | |
Polyclonal | |
Unconjugated | |
OR2W3 | |
Olfactory receptor 2W3; olfactory receptor 2W8; olfactory receptor family 2 subfamily W member 3; olfactory receptor OR1-49; olfactory receptor, family 2, subfamily W, member 3 pseudogene; olfactory receptor, family 2, subfamily W, member 8 pseudogene; OR2W3; OR2W3P; OR2W8P; OST718 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
343171 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q7Z3T1 | |
OR2W3 | |
Recombinant protein corresponding to Human OR2W3. Recombinant protein control fragment (Product #RP-100620). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction