Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR4D2 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548765
Description
Odorant receptor.Specifications
OR4D2 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P58180 | |
OR4D2 | |
KLH conjugated synthetic peptide between 211-239 amino acids from the C-terminal region of human OR4D2. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
OR4D2 | |
Olfactory receptor 4D2, B-lymphocyte membrane protein BC2009, Olfactory receptor OR17-24, OR4D2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
124538 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction