Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR4S2 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548971

Catalog No. PA548971


Add to cart

Description

Description

Odorant receptor.
Specifications

Specifications

OR4S2
Polyclonal
PBS with 0.09% sodium azide
Q8NH73
Olfactory receptor 4S2, Olfactory receptor OR11-137, OR4S2, OR4S2P
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
219431
Western Blot
Unconjugated
OR4S2
Liquid
OR4S2
KLH conjugated synthetic peptide between 206-233 amino acids from the C-terminal region of human OR4S2.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit