Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR51G1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548998
Description
OR51G1 Polyclonal Antibody for Western Blot
Odorant receptor.
Specifications
OR51G1 | |
Polyclonal | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8NGK1 | |
OR51G1 | |
KLH conjugated synthetic peptide between 287-316 amino acids from the C-terminal region of human OR51G1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
OR51G1 | |
Olfactory receptor 51G1; olfactory receptor 51G3; olfactory receptor family 51 subfamily G member 1 (gene/pseudogene); olfactory receptor OR11-29; olfactory receptor, family 51, subfamily G, member 1; OR11-29; OR51G1; OR51G3P | |
Rabbit | |
Antigen Affinity Chromatography, Protein A | |
RUO | |
79324 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction