Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR51M1 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548970

Catalog No. PA548970

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Odorant receptor.
Specifications

Specifications

OR51M1
Polyclonal
PBS with 0.09% sodium azide
Q9H341
Olfactory receptor 51M1, Odorant receptor HOR5′beta7, Olfactory receptor OR11-40, OR51M1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
390059
Western Blot
Unconjugated
OR51M1
Liquid
OR51M1
KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human OR51M1.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit