Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

OR52N5 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548362

Catalog No. PA548362


Add to cart

Description

Description

Odorant receptor.
Specifications

Specifications

OR52N5
Polyclonal
PBS with 0.09% sodium azide
Q8NH56
Olfactory receptor 52N5, Olfactory receptor OR11-62, OR52N5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
390075
Western Blot
Unconjugated
OR52N5
Liquid
OR52N5
KLH conjugated synthetic peptide between 292-321 amino acids from the C-terminal region of human OR52N5.
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit