Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR7D4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570793
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
OR7D4 is a protein coding gene.
Specifications
OR7D4 | |
Polyclonal | |
Unconjugated | |
OR7D4 | |
hg105; odorant receptor family 7 subfamily D member 4 RT; Odorant receptor family subfamily D member 4RT; Olfactory receptor 7D4; olfactory receptor family 7 subfamily D member 4; olfactory receptor OR19-7; olfactory receptor, family 7, subfamily D, member 4 pseudogene; OR19-7; OR19B; OR19-B; OR7D4; OR7D4P | |
Rabbit | |
Affinity Chromatography | |
RUO | |
125958 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8NG98 | |
OR7D4 | |
synthetic peptide directed towards the C-terminal region of human OR7D4. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction