Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ OR8A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA549225
Description
OR8A1 Polyclonal Antibody for Western Blot
Odorant receptor.
Specifications
OR8A1 | |
Polyclonal | |
Unconjugated | |
OR8A1 | |
Olfactory receptor 8A1; olfactory receptor family 8 subfamily A member 1; olfactory receptor OR11-318; olfactory receptor, family 8, subfamily A, member 1; OR11-318; OR8A1; OST025 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
390275 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8NGG7 | |
OR8A1 | |
KLH conjugated synthetic peptide between 10-39 amino acids from the N-terminal region of human OR8A1. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction