Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OR8B12 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA548712

Catalog No. PA548712


Only null left
Add to Cart

Description

Description

OR8B12 Polyclonal Antibody for Western Blot

Odorant receptor.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OR8B12
Polyclonal
PBS with 0.09% sodium azide; pH 7.4
Q8NGG6
OR8B12
KLH conjugated synthetic peptide between 279-307 amino acids from the C-terminal region of human OR8B12.
400 μL
Primary
Human
Antibody
IgG
Western Blot
Unconjugated
OR8B12
olfactory receptor 8B12; olfactory receptor family 8 subfamily B member 12; olfactory receptor OR11-317; olfactory receptor, family 8, subfamily B, member 12; OR11-317; OR8B12
Rabbit
Antigen affinity chromatography, Protein A
RUO
219858
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.