Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR8H1 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | OR8H1 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB124555
|
Novus Biologicals
NBP309827100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
OR8H1 Polyclonal specifically detects OR8H1 in Human samples. It is validated for Western Blot.Specifications
OR8H1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
olfactory receptor 8H1, Olfactory receptor OR11-180, olfactory receptor, family 8, subfamily H, member 1, OR11-180 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR8H1 (NP_001005199). Peptide sequence YGTMIFTYLKPRKSYSLGRDQVASVFYTIVIPMLNPLIYSLRNKEVKNAL | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
219469 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title