Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OR9G9 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | OR9G9 |
---|---|
Dilution | Western Blot 1.0 ug/ml |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB124579
|
Novus Biologicals
NBP309839100UL |
100 μg |
Each of 1 for $482.50
|
|
|||||
Description
OR9G9 Polyclonal specifically detects OR9G9 in Human samples. It is validated for Western Blot.Specifications
OR9G9 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
olfactory receptor 9G9, olfactory receptor, family 9, subfamily G, member 9 | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human OR9G9 (NP_001013376). Peptide sequence YIYALPRSSYSFDMDKIVSTFYTEVLPMLNPMIYSLRNKDVKEALKKLLP | |
Affinity purified |
Western Blot 1.0 ug/ml | |
Polyclonal | |
Purified | |
RUO | |
PBS buffer, 2% sucrose | |
504191 | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title