Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORAI1 (extracellular) Monoclonal Antibody (3F11/D10/B9), Invitrogen™

Catalog No. PIMA527545
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIMA527545 100 μg
1 options

Catalog No. PIMA527545

Supplier: Thermo Scientific MA527545

Antibody Cocktail

Product is shipped at room temperature as a lyophilized powder and should be stored at -20C upon receipt. Reconstitute in 25 uL OR 100 uL DDW, depending on sample size.

ORAI1 is a plasma membrane protein essential for store-operated calcium entry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ORAI1 (extracellular)
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Cocktail
Clone 3F11/D10/B9
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.05% sodium azide; pH 7.4
Gene ORAI1
Gene Accession No. Q96D31
Gene Alias CRACM1, TMEM142A
Gene Symbols ORAI1
Host Species Mouse
Immunogen Synthetic peptide mapping to an extracellular sequence of human Orai1
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84876
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.