3M™ Orange County Choppers™ Eyewear
Impact-resistant polycarbonate lenses absorb 99.9% of UV rays
Manufacturer: 3M™ 11734-00000-10
 View all options for this product
Catalog No. 18-120-261
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Certifications/Compliance 200, 300, 400, 700, 800, 1100, and 1200 series meet ANSI Z87.1-2003 standards. 100 Series meets ANSI Z87.1-2003 and CSA Z94.3-2007 standards.
    Includes Microfiber bag and lanyard
    Lens Material Polycarbonate
    Lens Tint I/O Mirror
    Frame Color Metalik Gun Metal Gray Frame
    Lens Coating Hardcoated
    100, 200, 300, and 400 Series
    • Durable, lightweight wraparound aluminum frames
    • Spring-hinge temples with soft tips and adjustable nosebridge
    • OCC logo on temple
    700 Series
    • Lightweight, sporty half-frame with soft-tip temples
    • OCC logo on temple
    800 Series
    • Sleek, frameless design
    • Cross-member black and silver temples
    • OCC logo on lens
    1100 Series
    Wraparound frame with wide temples
    Soft, adjustable nosebridge
    OCC logo on temple
    1200 Series
    • Lightweight, half-frame
    • Soft, adjustable nosebridge
    • OCC logo on temple
  • Description & Specifications
Related Products
Product Content Feedback