Learn More
MCR Safety Orange Aramid Fiber/Orange Nitrile Foam
Orange Aramid Fiber/Orange Nitrile Foam features a patent-protected, high visibility orange aramid fiber yarn. The high visibility orange enhances safety awareness.
Manufacturer:  MCR Safety 9178NFOM
 View more versions of this product
Catalog No. 19-812-636
Add to cart
Description
Description
  • Patent-protected, high visibility orange DuPont™ Kevlar™ Aramid fibers® engineered yarn
  • High Visibility orange enhances safety awareness
  • Light weight seamless shell provides dexterity and comfort
  • Orange nitrile foam coating allows for tacky grip and sense of touch
  • Touch screen friendly
Specifications
Specifications
Nitrile
Knit Wrist
Manufacturing
Medium
10
Powder-free
Non-sterile
4
Multi Colored
Dupont™ Kevlar™ aramid fibers
1 Pair
Glove
Orange
30.5