Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Orange Aramid Fiber/Orange Nitrile Foam

Orange Aramid Fiber/Orange Nitrile Foam features a patent-protected, high visibility orange aramid fiber yarn. The high visibility orange enhances safety awareness.

$23.37

Specifications

Color Orange
For Use With (Application) Manufacturing
Glove Cuff Style Knit Wrist
Material Dupont™ Kevlar™ aramid fibers
Powdered Powder-free
View More Specs

Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-812-634 View Documents MCR Safety
9178NFOXS
X-Small Pair for $23.37
Add to cart
 
19-812-635 View Documents MCR Safety
9178NFOS
Small Pair for $23.37
Add to cart
 
19-812-636 View Documents MCR Safety
9178NFOM
Medium Pair for $23.37
Add to cart
 
19-812-637 View Documents MCR Safety
9178NFOL
Large Pair for $23.37
Add to cart
 
19-812-638 View Documents MCR Safety
9178NFOXL
X-Large Pair for $23.37
Add to cart
 
19-812-639 View Documents MCR Safety
9178NFOXXL
2X-Large Pair for $23.37
Add to cart
 
Description

Description

  • Patent-protected, high visibility orange DuPont™ Kevlar™ aramid fibers engineered yarn
  • High Visibility orange enhances safety awareness
  • Light weight seamless shell provides dexterity and comfort
  • Orange nitrile foam coating allows for tacky grip and sense of touch
  • Touch screen friendly
Specifications

Specifications

Orange
Knit Wrist
Powder-free
Non-sterile
Manufacturing
Dupont™ Kevlar™ aramid fibers
1 Pair
Glove
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit