Learn More
Invitrogen™ ORAOV1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier:  Invitrogen™ PA551030
Catalog No. PA551030
Only null left
Add to Cart
Description
Description
ORAOV1 Polyclonal Antibody detects endogenous levels of ORAOV1 protein.
ORAOV1 is a protein coding gene. The complex LTO1:YAE1 functions as a target specific adapter that probably recruits apo-ABCE1 to the cytosolic iron-sulfur protein assembly (CIA) complex machinery. May be required for biogenesis of the large ribosomal subunit and initiation of translation. May play a role in the regulation of proline metabolism and ROS production.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ORAOV1
Polyclonal
Unconjugated
ORAOV1
2210010N10Rik; Lto1; oral cancer overexpressed 1; oral cancer overexpressed protein 1-A; oral cancer-overexpressed protein 1; Oral cancer-overexpressed protein 1 homolog; Oraov1; Protein LTO1 homolog; TAOS1; Tumor-amplified and overexpressed sequence 1
Rabbit
Antigen Affinity Chromatography
RUO
220064
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q8WV07
ORAOV1
Synthesized peptide derived from the Internal region of human ORAOV1 (aa 81-130).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.