Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
OraSure Technologies OraQuick™ HCV Antibody Test
HCV Testing anytime, anywhere made easy. The OraQuick™ HCV test is the only point-of-care Hepatitis C test on the market today, detecting HCV antibodies in fingerstick and venipuncture whole blood. Yielding accurate results in as little as 20 minutes.
Manufacturer:  OraSure Technologies 10010181
 View more versions of this product
Catalog No. 23-222-105
Add to cart
This item is not returnable. View return policy
Description
Description
  • The OraQuick™ HCV test is FDA approved for detecting HCV antibodies in fingerstick and venipuncture whole blood.
  • Our simple platform enables healthcare providers to deliver an accurate result in 20 minutes.
  • The OraQuick™ HCV Rapid Antibody Test is comprised of both a single-use test device and vial containing a pre-measured amount of a buffered developer solution.
  • The test consists of a sealed pouch with two separate compartments for each component.
  • The developer solution facilitates the capillary flow of the specimen into the device and onto the assay strip.
  • 25 Test Kit includes: 25 divided pouches each containing a test device, absorbent packet, and developer solution vial (each contains 0.75 mL of a buffered saline solution with an antimicrobial agent); 5 reusable test stands; 25 specimen collection loops; package insert.
  • 100 Test Kit includes: 100 divided pouches each containing a test device, absorbent packet, and developer solution vial (each contains 0.75 mL of a buffered saline solution with an antimicrobial agent); 5 reusable test stands; 25 specimen collection loops; package insert.
Specifications
Specifications
ISO 13485 Certified. FDA approved under PMA P080027. Inspected to 21 CFR 820 and MDSAP certification.
Yes
25 Tests
2°C to 30°C
Contains synthetic peptides and recombinant antigen from the core, NS3 and NS4 regions of the HCV genome.
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Cancel Submit