Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
OraSure Technologies OraQuick™ HCV Antibody Test

Click to view available options
Quantity:
100 Tests
25 Tests
Description
- The OraQuick™ HCV test is FDA approved for detecting HCV antibodies in fingerstick and venipuncture whole blood.
- Our simple platform enables healthcare providers to deliver an accurate result in 20 minutes.
- The OraQuick™ HCV Rapid Antibody Test is comprised of both a single-use test device and vial containing a pre-measured amount of a buffered developer solution.
- The test consists of a sealed pouch with two separate compartments for each component.
- The developer solution facilitates the capillary flow of the specimen into the device and onto the assay strip.
- 25 Test Kit includes: 25 divided pouches each containing a test device, absorbent packet, and developer solution vial (each contains 0.75 mL of a buffered saline solution with an antimicrobial agent); 5 reusable test stands; 25 specimen collection loops; package insert.
- 100 Test Kit includes: 100 divided pouches each containing a test device, absorbent packet, and developer solution vial (each contains 0.75 mL of a buffered saline solution with an antimicrobial agent); 5 reusable test stands; 25 specimen collection loops; package insert.
Specifications
Specifications
Certifications/Compliance | ISO 13485 Certified. FDA approved under PMA P080027. Inspected to 21 CFR 820 and MDSAP certification. |
Content And Storage | 2°C to 30°C |
CE Marker | Yes |
Detectable Analytes | HCV |
Quantity | 25 Tests |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction