Learn More
MilliporeSigma™ Supelco™ ORBO™ 80 Coated Filter with Cassettes (unassembled)
Used for gas and vapor sampling
Supplier:  MilliporeSigma™ Supelco™ 20812U
Catalog No. 11-100-4064
Add to Cart
Description
Description
ORBO™ 80 Coated Filter, a glass fiber filter coated with 1-(2-pyridyl)piperazine, is generally used for sampling diisocyanates in air.
Specifications
Specifications
With cassettes (unassembled)
25/Pk.
Product Suggestions
Product Suggestions
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
ORBO is a trademark of Sigma-Aldrich Co. LLC