Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ORC5L Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548510

Catalog No. PA548510

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Component of the origin recognition complex (ORC) that binds origins of replication. DNA-binding is ATP-dependent. The specific DNA sequences that define origins of replication have not been identified yet. ORC is required to assemble the pre- replication complex necessary to initiate DNA replication.
Specifications

Specifications

ORC5L
Polyclonal
PBS with 0.09% sodium azide
Q9WUV0
Origin recognition complex subunit 5, ORC5, ORC5L
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
26429
Western Blot
Unconjugated
Orc5
Liquid
ORC5
KLH conjugated synthetic peptide between 227-256 amino acids from the Central region of human ORC5L.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit