Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Orcinol Monohydrate, 99%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
6153-39-5
C7H8O2·H2O
142.15
MFCD00149092
NBKPNAMTHBIMLA-UHFFFAOYSA-N
5-methylbenzene-1,3-diol hydrate,orcinol monohydrate,5-methylresorcinol monohydrate,3,5-dihydroxytoluene monohydrate,orcinol hydrate,resorcinol, 5-methyl-, monohydrate,1,3-benzenediol, 5-methyl-, monohydrate,1,3-dihydroxy-5-methylbenzene monohydrate,acmc-209mtl,5-methylresorcinol hydrate
3083941
5-methylbenzene-1,3-diol;hydrate
CC1=CC(=CC(=C1)O)O.O
6153-39-5
C7H8O2·H2O
142.15
MFCD00149092
NBKPNAMTHBIMLA-UHFFFAOYSA-N
5-methylbenzene-1,3-diol hydrate,orcinol monohydrate,5-methylresorcinol monohydrate,3,5-dihydroxytoluene monohydrate,orcinol hydrate,resorcinol, 5-methyl-, monohydrate,1,3-benzenediol, 5-methyl-, monohydrate,1,3-dihydroxy-5-methylbenzene monohydrate,acmc-209mtl,5-methylresorcinol hydrate
3083941
5-methylbenzene-1,3-diol;hydrate
CC1=CC(=CC(=C1)O)O.O
Infrared Spectrum Authentic
Melting Point 56.0°C to 61.0°C
Water 10 to 14% (K.F.)
Color White
Quantity 100g
Boiling Point 290.0°C
Packaging Glass bottle
Assay Percent Range 99%
Beilstein 06, 882
Merck Index 14, 6864
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and ether, freely soluble in benzene, slightly soluble in chloroform and carbon, disulfide
Formula Weight 142.15
Physical Form Powder, Crystals or Flakes
Percent Purity 99%
Chemical Name or Material Orcinol monohydrate
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.